Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nle1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145431.2 | NP_663406.2 | 11 - 12 | 1408 | 96 | |
GenBank mRNA | AI852473.1 | - | 2 - 3 | 96 | ||
AK141230.1 | - | 11 - 12 | 1383 | 96 | ||
AK170853.1 | - | 11 - 12 | 1381 | 96 | ||
BC018399.1 | - | 11 - 12 | 1399 | 3491925 | 96 | |
BC035421.1 | - | 11 - 12 | 1399 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |