Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fsd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172904.2 | NP_766492.2 | 4 - 5 | 1051 | 66 | |
GenBank mRNA | AK028142.1 | - | 4 - 5 | 1043 | 66 | |
AK036674.1 | - | 1017 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |