Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162979.1 | NP_001156451.1 | 12 - 13 | 1840 | 124 | |
GenBank mRNA | AK014876.1 | - | 12 - 13 | 1823 | 124 | |
BC127049.1 | - | 12 - 13 | 1827 | 124 | ||
BC152307.1 | - | 12 - 13 | 1829 | 40062409 | 124 | |
BC152308.1 | - | 12 - 13 | 1829 | 40062416 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |