Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145806.1 | NP_001139278.1 | 4 - 5 | 610 | 84 | |
NM_130890.2 | NP_570960.2 | 4 - 5 | 610 | 84 | ||
GenBank mRNA | AB050202.1 | - | 4 - 5 | 606 | 84 | |
AB050203.1 | - | 4 - 5 | 606 | 84 | ||
AB061518.1 | - | 4 - 5 | 556 | 84 | ||
AB061519.1 | - | 4 - 5 | 556 | 84 | ||
AK132449.1 | - | 4 - 5 | 611 | 84 | ||
BC125504.1 | - | 4 - 5 | 582 | 40130019 | 84 | |
BC125508.1 | - | 4 - 5 | 582 | 40130023 | 84 | |
BC145055.1 | - | 4 - 5 | 582 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |