Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293806.1 | NP_001280735.1 | 16 - 17 | 3041 | 77 | |
NM_027777.2 | NP_082053.1 | 15 - 16 | 2921 | 77 | ||
GenBank mRNA | AK017309.1 | - | 3 - 4 | 338 | 77 | |
BC090845.1 | - | 15 - 16 | 2921 | 6400629 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |