Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK034856.1 | - | 2 - 3 | 316 | 76 | |
AK085362.1 | - | 2 - 3 | 353 | 76 | ||
AK086462.1 | - | 2 - 3 | 346 | 76 | ||
BC054120.1 | - | 2 - 3 | 299 | 6506658 | 76 | |
CD348032.1 | - | 2 - 3 | 443 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |