Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Susd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144796.4 | NP_659045.2 | 1 - 2 | 281 | 67 | |
GenBank mRNA | AK035779.1 | - | 1 - 2 | 281 | 67 | |
AK088629.1 | - | 1 - 2 | 194 | 67 | ||
BC021842.1 | - | 1 - 2 | 291 | 5135798 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |