Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcoln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134160.1 | NP_598921.1 | 9 - 10 | 1193 | 64 | |
GenBank mRNA | AF475086.1 | - | 9 - 10 | 1193 | 64 | |
AK030819.1 | - | 9 - 10 | 1313 | 64 | ||
AK033008.1 | - | 9 - 10 | 1307 | 64 | ||
AK035029.1 | - | 9 - 10 | 1314 | 64 | ||
AY083531.1 | - | 9 - 10 | 1191 | 64 | ||
BC106856.2 | - | 9 - 10 | 1193 | 40047039 | 64 | |
BC106857.2 | - | 9 - 10 | 1193 | 40047042 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |