Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec24b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207209.2 | NP_997092.1 | 19 - 20 | 3445 | 66 | |
GenBank mRNA | AK150076.1 | - | 18 - 19 | 2522 | 66 | |
BC046776.1 | - | 19 - 20 | 3445 | 5706007 | 66 | |
BC057355.1 | - | 19 - 20 | 3305 | 6833698 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |