Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022019.5 | NP_071302.2 | 2 - 3 | 958 | 83 | |
GenBank mRNA | AB037908.1 | - | 2 - 3 | 964 | 83 | |
AK011995.1 | - | 2 - 3 | 942 | 83 | ||
AK035293.1 | - | 2 - 3 | 956 | 83 | ||
AK050528.1 | - | 2 - 3 | 956 | 83 | ||
AK088024.1 | - | 2 - 3 | 957 | 83 | ||
AK088186.1 | - | 2 - 3 | 959 | 83 | ||
AK088357.1 | - | 2 - 3 | 954 | 83 | ||
AK142568.1 | - | 2 - 3 | 960 | 83 | ||
BC025066.1 | - | 2 - 3 | 976 | 3495358 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |