Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Homer2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164086.1 | NP_001157558.1 | 7 - 8 | 918 | 84 | |
NM_001164087.1 | NP_001157559.1 | 7 - 8 | 934 | 84 | ||
NM_011983.2 | NP_036113.1 | 7 - 8 | 951 | 84 | ||
GenBank mRNA | AB017136.1 | - | 7 - 8 | 944 | 84 | |
AF093259.1 | - | 7 - 8 | 823 | 84 | ||
AF093260.1 | - | 7 - 8 | 856 | 84 | ||
AK158673.1 | - | 1020 | 84 | |||
BC038314.1 | - | 3 - 4 | 592 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |