Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Styx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019637.3 | NP_062611.2 | 5 - 6 | 592 | 160 | |
GenBank mRNA | AK002822.1 | - | 534 | 160 | ||
AK030122.1 | - | 5 - 6 | 592 | 160 | ||
AK042741.1 | - | 6 - 7 | 647 | 160 | ||
AK046970.1 | - | 5 - 6 | 569 | 160 | ||
AK154136.1 | - | 5 - 6 | 687 | 160 | ||
AK166043.1 | - | 5 - 6 | 575 | 160 | ||
BC038608.1 | - | 5 - 6 | 542 | 5012303 | 160 | |
U34973.1 | - | 5 - 6 | 560 | 160 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |