Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110142.1 | NP_001103612.1 | 15 - 16 | 2093 | 140 | |
NM_028288.5 | NP_082564.3 | 15 - 16 | 2034 | 140 | ||
GenBank mRNA | AB093259.1 | - | 11 - 12 | 1651 | 140 | |
AK012410.1 | - | 13 - 14 | 1797 | 140 | ||
AK032701.1 | - | 15 - 16 | 2035 | 140 | ||
AK160998.1 | - | 13 - 14 | 1804 | 140 | ||
AK170632.1 | - | 12 - 13 | 2198 | 140 | ||
AY330868.1 | - | 15 - 16 | 2035 | 140 | ||
BC004026.1 | - | 15 - 16 | 2082 | 3494708 | 140 | |
BC010347.1 | - | 2 - 3 | 220 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |