Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136057.2 | NP_001129529.1 | 11 - 12 | 1342 | 69 | |
NM_001146183.1 | NP_001139655.1 | 11 - 12 | 1319 | 69 | ||
NM_009947.3 | NP_034077.1 | 11 - 12 | 1145 | 69 | ||
GenBank mRNA | AB008893.1 | - | 11 - 12 | 1145 | 69 | |
AK134548.1 | - | 11 - 12 | 1164 | 69 | ||
AK144410.1 | - | 11 - 12 | 1171 | 69 | ||
BC050766.1 | - | 11 - 12 | 1167 | 6591063 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |