Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700029809.mm.1 |
NM_001038643.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029809.mm.1 |
NM_001038643.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029809.mm.1 |
NM_023908.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029809.mm.1 |
NM_023908.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029809.mm.1 |
TC0700003517.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029812.mm.1 |
NM_001038643.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029812.mm.1 |
NM_001038643.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029812.mm.1 |
NM_023908.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029812.mm.1 |
NM_023908.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029812.mm.1 |
TC0700003517.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029805.mm.1 |
NM_001038643.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029805.mm.1 |
NM_001038643.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029805.mm.1 |
NM_023908.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029805.mm.1 |
NM_023908.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029805.mm.1 |
TC0700003517.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029808.mm.1 |
NM_001038643.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029808.mm.1 |
NM_001038643.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029808.mm.1 |
NM_023908.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029808.mm.1 |
NM_023908.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029808.mm.1 |
TC0700003517.mm.1 |
MTA TC |