Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026231.2 | NP_080507.2 | 10 - 11 | 1885 | 119 | |
GenBank mRNA | AK013967.1 | - | 7 - 8 | 737 | 119 | |
AK017059.1 | - | 8 - 9 | 1160 | 119 | ||
AK020021.1 | - | 9 - 10 | 1495 | 119 | ||
AK029724.1 | - | 10 - 11 | 1886 | 119 | ||
AK033409.1 | - | 10 - 11 | 1924 | 119 | ||
BC047136.1 | - | 10 - 11 | 1885 | 6311866 | 119 | |
BC061240.1 | - | 10 - 11 | 1881 | 6743615 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |