Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177381.3 | NP_796355.2 | 2 - 3 | 377 | 100 | |
GenBank mRNA | AK140362.1 | - | 2 - 3 | 377 | 100 | |
BC038030.1 | - | 3 - 4 | 388 | 4020725 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |