Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdadc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168535.1 | NP_001162007.1 | 6 - 7 | 1175 | 74 | |
NM_001168536.1 | NP_001162008.1 | 6 - 7 | 1175 | 74 | ||
NM_001168537.1 | NP_001162009.1 | 6 - 7 | 1175 | 74 | ||
NM_001168538.1 | NP_001162010.1 | 6 - 7 | 1175 | 74 | ||
NM_027986.3 | NP_082262.1 | 6 - 7 | 1175 | 74 | ||
GenBank mRNA | AK009280.1 | - | 6 - 7 | 1167 | 74 | |
AK032600.1 | - | 6 - 7 | 1154 | 74 | ||
AK032813.1 | - | 6 - 7 | 1161 | 74 | ||
AK038926.1 | - | 6 - 7 | 1155 | 74 | ||
AK039148.1 | - | 6 - 7 | 1163 | 74 | ||
BC004588.1 | - | 2 - 3 | 190 | 74 | ||
BC006901.1 | - | 6 - 7 | 1137 | 3600168 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |