Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chmp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134078.4 | NP_598839.2 | 1 - 2 | 383 | 94 | |
GenBank mRNA | BC024115.1 | - | 1 - 2 | 371 | 5100379 | 94 |
BC033365.1 | - | 1 - 2 | 354 | 5367653 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |