Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oxa1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC027191.1 | - | 3 - 4 | 341 | 4223847 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |