Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sertad4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177794.1 | NP_001171265.1 | 2 - 3 | 242 | 96 | |
NM_198247.2 | NP_937890.2 | 2 - 3 | 497 | 96 | ||
GenBank mRNA | BC060251.1 | - | 2 - 3 | 466 | 5708574 | 96 |
BX514346.1 | - | 2 - 3 | 243 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |