Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhx8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010713.2 | NP_034843.2 | 1 - 2 | 289 | 78 | |
GenBank mRNA | AK160121.1 | - | 1 - 2 | 290 | 78 | |
AK165733.1 | - | 1 - 2 | 283 | 78 | ||
BC125281.1 | - | 1 - 2 | 198 | 40129796 | 78 | |
BC125283.1 | - | 1 - 2 | 198 | 40129798 | 78 | |
BC144768.1 | - | 1 - 2 | 198 | 9053316 | 78 | |
D49658.1 | - | 1 - 2 | 258 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |