Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080435.1 | NP_536683.1 | 7 - 8 | 1048 | 76 | |
GenBank mRNA | AF442771.1 | - | 7 - 8 | 1048 | 76 | |
AK141767.1 | - | 7 - 8 | 1119 | 76 | ||
AK161880.1 | - | 7 - 8 | 1067 | 76 | ||
AK220443.1 | - | 5 - 6 | 1037 | 76 | ||
BC015299.1 | - | 7 - 8 | 1048 | 4236007 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |