Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028665.4 | NP_082941.2 | 8 - 9 | 1969 | 86 | |
GenBank mRNA | AK016847.1 | - | 3 - 4 | 250 | 86 | |
AK029511.1 | - | 8 - 9 | 1971 | 86 | ||
AK029888.1 | - | 8 - 9 | 1930 | 86 | ||
AK133339.1 | - | 1930 | 86 | |||
BC030885.1 | - | 8 - 9 | 1598 | 86 | ||
BC100433.1 | - | 8 - 9 | 2005 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |