Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033314.3 | NP_001028486.1 | 4 - 5 | 667 | 124 | |
GenBank mRNA | AK146297.1 | - | 4 - 5 | 668 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |