Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030559.3 | NP_085036.3 | 8 - 9 | 883 | 88 | |
GenBank mRNA | AB056721.1 | - | 8 - 9 | 841 | 88 | |
AK049872.1 | - | 8 - 9 | 840 | 88 | ||
AK162984.1 | - | 7 - 8 | 990 | 88 | ||
BC025626.1 | - | 8 - 9 | 849 | 5320572 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |