Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009322.3 | NP_033348.2 | 2 - 3 | 1099 | 86 | |
GenBank mRNA | BC052737.1 | - | 2 - 3 | 1099 | 6825394 | 86 |
BC058399.1 | - | 2 - 3 | 1003 | 6826831 | 86 | |
U49251.1 | - | 2 - 3 | 939 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |