Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Leng8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172736.3 | NP_766324.1 | 12 - 13 | 1987 | 78 | |
GenBank mRNA | AK034310.1 | - | 12 - 13 | 1992 | 78 | |
AK170629.1 | - | 12 - 13 | 1995 | 78 | ||
BC034448.1 | - | 4 - 5 | 711 | 78 | ||
BC041775.1 | - | 11 - 12 | 2492 | 78 | ||
BC042658.1 | - | 12 - 13 | 1975 | 5365041 | 78 | |
BC066768.1 | - | 12 - 13 | 1978 | 6407146 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |