Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sel1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033296.2 | NP_001028468.1 | 8 - 9 | 817 | 135 | |
GenBank mRNA | AK016809.1 | - | 8 - 9 | 834 | 135 | |
AK132650.1 | - | 8 - 9 | 818 | 135 | ||
BC150948.1 | - | 8 - 9 | 789 | 9087859 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |