Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp8b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080944.3 | NP_001074413.2 | 8 - 9 | 613 | 72 | |
GenBank mRNA | AK040903.1 | - | 9 - 10 | 782 | 72 | |
AK086607.1 | - | 8 - 9 | 647 | 72 | ||
AK155448.1 | - | 8 - 9 | 624 | 72 | ||
AK169967.1 | - | 8 - 9 | 636 | 72 | ||
AK173307.1 | - | 8 - 9 | 613 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Mus musculus Probe spans exons |