Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hhipl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044380.1 | NP_001037845.1 | 7 - 8 | 1896 | 83 | |
GenBank mRNA | AK129450.1 | - | 1291 | 83 | ||
AK147996.1 | - | 1563 | 83 | |||
AK166269.1 | - | 1792 | 83 | |||
BC057108.1 | - | 889 | 83 | |||
BC113163.1 | - | 7 - 8 | 1896 | 40060054 | 83 | |
BC113773.1 | - | 7 - 8 | 1892 | 40060051 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |