Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmpca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173180.3 | NP_775272.1 | 2 - 3 | 282 | 138 | |
GenBank mRNA | AK004549.1 | - | 2 - 3 | 282 | 138 | |
AK032081.1 | - | 2 - 3 | 282 | 138 | ||
AK075965.1 | - | 2 - 3 | 283 | 138 | ||
AK077218.1 | - | 2 - 3 | 283 | 138 | ||
AK147110.1 | - | 2 - 3 | 285 | 138 | ||
AK150118.1 | - | 2 - 3 | 283 | 138 | ||
AK158990.1 | - | 2 - 3 | 250 | 138 | ||
AK161291.1 | - | 2 - 3 | 283 | 138 | ||
AK169325.1 | - | 2 - 3 | 277 | 138 | ||
AK169342.1 | - | 2 - 3 | 277 | 138 | ||
BC010810.1 | - | 2 - 3 | 267 | 4241697 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |