Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp6r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172894.2 | NP_766482.2 | 19 - 20 | 2719 | 86 | |
GenBank mRNA | AK075825.1 | - | 3 - 4 | 431 | 86 | |
AK122450.1 | - | 15 - 16 | 3823 | 86 | ||
BC053076.1 | - | 19 - 20 | 2719 | 5698212 | 86 | |
BC141039.1 | - | 18 - 19 | 2266 | 9055651 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |