Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027000.4 | NP_081276.2 | 8 - 9 | 922 | 152 | |
GenBank mRNA | AF348208.1 | - | 8 - 9 | 929 | 152 | |
AK011580.1 | - | 8 - 9 | 933 | 152 | ||
AK141852.1 | - | 8 - 9 | 932 | 152 | ||
AK164330.1 | - | 8 - 9 | 923 | 152 | ||
BC005514.1 | - | 8 - 9 | 903 | 152 | ||
BC025431.1 | - | 8 - 9 | 927 | 5007174 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |