Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rxra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290481.1 | NP_001277410.1 | 7 - 8 | 1111 | 73 | |
NM_001290482.1 | NP_001277411.1 | 7 - 8 | 1148 | 73 | ||
NM_011305.3 | NP_035435.1 | 7 - 8 | 1290 | 73 | ||
GenBank mRNA | AK031034.1 | - | 7 - 8 | 1159 | 73 | |
AK144675.1 | - | 7 - 8 | 1292 | 73 | ||
BC138800.1 | - | 8 - 9 | 1141 | 8861822 | 73 | |
BC138802.1 | - | 8 - 9 | 1141 | 8861824 | 73 | |
M84817.1 | - | 8 - 9 | 1153 | 73 | ||
U77683.1 | - | 7 - 8 | 1121 | 73 | ||
U77684.1 | - | 7 - 8 | 1151 | 73 | ||
X66223.1 | - | 7 - 8 | 1235 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Nuclear Receptor Mammalian Gene Collection Mus musculus Probe spans exons |