Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025358.2 | NP_001020529.2 | 5 - 6 | 971 | 77 | |
NM_001025359.2 | NP_001020530.2 | 5 - 6 | 971 | 77 | ||
NM_001025360.2 | NP_001020531.2 | 5 - 6 | 990 | 77 | ||
NM_001025361.2 | NP_001020532.2 | 5 - 6 | 971 | 77 | ||
NM_001025362.2 | NP_001020533.2 | 5 - 6 | 971 | 77 | ||
NM_001025363.2 | NP_001020534.2 | 5 - 6 | 971 | 77 | ||
NM_001081959.1 | NP_001075428.1 | 5 - 6 | 990 | 77 | ||
NM_008450.2 | NP_032476.2 | 5 - 6 | 990 | 77 | ||
GenBank mRNA | AF055665.1 | - | 5 - 6 | 883 | 77 | |
AK031309.1 | - | 5 - 6 | 962 | 77 | ||
AY753300.1 | - | 4 - 5 | 795 | 77 | ||
BC055744.1 | - | 5 - 6 | 826 | 6415280 | 77 | |
BG094009.1 | - | 1 - 2 | 201 | 77 | ||
BU922789.1 | - | 1 - 2 | 42 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |