Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pygb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153781.1 | NP_722476.1 | 3 - 4 | 500 | 70 | |
GenBank mRNA | AK032769.1 | - | 3 - 4 | 513 | 70 | |
AK035673.1 | - | 3 - 4 | 71 | |||
AK134667.1 | - | 3 - 4 | 510 | 70 | ||
AK147763.1 | - | 3 - 4 | 532 | 70 | ||
AK169052.1 | - | 3 - 4 | 535 | 70 | ||
AK169631.1 | - | 3 - 4 | 514 | 70 | ||
BC031457.1 | - | 3 - 4 | 508 | 70 | ||
BC035283.1 | - | 3 - 4 | 500 | 4947455 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |