Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024465.3 | NP_077785.2 | 8 - 9 | 972 | 93 | |
GenBank mRNA | AK018261.1 | - | 8 - 9 | 906 | 93 | |
AK160126.1 | - | 8 - 9 | 972 | 93 | ||
AK178612.1 | - | 1 - 2 | 42 | 93 | ||
AK202769.1 | - | 1 - 2 | 42 | 93 | ||
AK216141.1 | - | 1 - 2 | 42 | 93 | ||
AK217770.1 | - | 1 - 2 | 42 | 93 | ||
AK219355.1 | - | 2 - 3 | 127 | 93 | ||
BC002138.1 | - | 8 - 9 | 941 | 93 | ||
BC002263.1 | - | 4 - 5 | 275 | 3495118 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |