Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301341.1 | NP_001288270.1 | 3 - 4 | 243 | 109 | |
GenBank mRNA | AK078777.1 | - | 3 - 4 | 244 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis Mus musculus Probe spans exons |