Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcm10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027290.3 | NP_081566.2 | 16 - 17 | 2440 | 72 | |
GenBank mRNA | AK030780.1 | - | 16 - 17 | 2385 | 72 | |
BC023236.1 | - | 8 - 9 | 1302 | 72 | ||
BC039663.1 | - | 2 - 3 | 289 | 72 | ||
BC064314.1 | - | 16 - 17 | 2803 | 72 | ||
BC094576.1 | - | 16 - 17 | 2440 | 6853824 | 72 | |
BC120687.1 | - | 15 - 16 | 2349 | 40129610 | 72 | |
BC120689.1 | - | 15 - 16 | 2349 | 40129612 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |