Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Optn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181848.4 | NP_862896.1 | 12 - 13 | 1588 | 78 | |
GenBank mRNA | AK015354.1 | - | 7 - 8 | 940 | 78 | |
AY071834.1 | - | 12 - 13 | 1695 | 78 | ||
BC024755.1 | - | 5 - 6 | 1474 | 78 | ||
BC061185.1 | - | 12 - 13 | 1588 | 30314331 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |