Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dab2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001602.2 | NP_001001602.2 | 7 - 8 | 1756 | 81 | |
NM_001114124.2 | NP_001107596.1 | 9 - 10 | 1880 | 81 | ||
NM_001114125.1 | NP_001107597.1 | 9 - 10 | 1732 | 81 | ||
NM_001290637.1 | NP_001277566.1 | 9 - 10 | 1787 | 81 | ||
NM_001290639.1 | NP_001277568.1 | 7 - 8 | 1415 | 81 | ||
NM_001290640.1 | NP_001277569.1 | 7 - 8 | 1720 | 81 | ||
NM_001290641.1 | NP_001277570.1 | 9 - 10 | 2013 | 81 | ||
GenBank mRNA | AK122548.1 | - | 4 - 5 | 720 | 81 | |
AK147464.1 | - | 9 - 10 | 1812 | 81 | ||
AK147593.1 | - | 9 - 10 | 2014 | 81 | ||
AK155296.1 | - | 9 - 10 | 1788 | 81 | ||
AY305656.1 | - | 7 - 8 | 1473 | 81 | ||
AY305657.1 | - | 7 - 8 | 1756 | 81 | ||
AY305658.1 | - | 7 - 8 | 1415 | 81 | ||
BC094373.1 | - | 4 - 5 | 577 | 81 | ||
BC118530.1 | - | 7 - 8 | 1381 | 40098202 | 81 | |
DQ473307.1 | - | 9 - 10 | 1617 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |