Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcp11l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177190.5 | NP_796164.1 | 4 - 5 | 618 | 73 | |
GenBank mRNA | AK047525.1 | - | 3 - 4 | 444 | 73 | |
AK082024.1 | - | 4 - 5 | 620 | 73 | ||
BC116926.1 | - | 3 - 4 | 439 | 40126245 | 73 | |
BC116932.1 | - | 3 - 4 | 439 | 40126251 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |