Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ell3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145973.2 | NP_666085.2 | 7 - 8 | 891 | 112 | |
GenBank mRNA | AK139676.1 | - | 8 - 9 | 1317 | 112 | |
BC045151.1 | - | 7 - 8 | 891 | 6313358 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |