Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011580.3 | NP_035710.2 | 12 - 13 | 2112 | 84 | |
GenBank mRNA | AK080686.1 | - | 12 - 13 | 2147 | 84 | |
AK163092.1 | - | 12 - 13 | 2139 | 84 | ||
BC050917.1 | - | 12 - 13 | 2112 | 6405710 | 84 | |
M87276.1 | - | 12 - 13 | 2138 | 84 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | AK089315.1 | - | 2278 | Chr. - - [-] on Build | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |