Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trip11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028446.1 | NP_082722.1 | 14 - 15 | 5388 | 90 | |
GenBank mRNA | BC044791.1 | - | 3 - 4 | 500 | 90 | |
BC158098.1 | - | 14 - 15 | 5484 | 9088179 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |