Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200009797.mm.1 |
NM_021398.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009797.mm.1 |
NM_021398.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009797.mm.1 |
TC0200001226.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200009801.mm.1 |
NM_021398.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009801.mm.1 |
NM_021398.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009801.mm.1 |
TC0200001226.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200009802.mm.1 |
NM_021398.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009802.mm.1 |
NM_021398.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009802.mm.1 |
TC0200001226.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200009800.mm.1 |
NM_021398.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009800.mm.1 |
NM_021398.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009800.mm.1 |
TC0200001226.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200009796.mm.1 |
NM_021398.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009796.mm.1 |
NM_021398.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009796.mm.1 |
TC0200001226.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200009803.mm.1 |
NM_021398.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009803.mm.1 |
NM_021398.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200009803.mm.1 |
TC0200001226.mm.1 |
MTA TC |