Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136081.2 | NP_001129553.1 | 10 - 11 | 1502 | 78 | |
NM_182990.4 | NP_892035.2 | 10 - 11 | 1631 | 78 | ||
GenBank mRNA | AK146446.1 | - | 10 - 11 | 1554 | 78 | |
AK146585.1 | - | 10 - 11 | 1551 | 78 | ||
AK146607.1 | - | 10 - 11 | 1534 | 78 | ||
AK146691.1 | - | 10 - 11 | 1533 | 78 | ||
AK147005.1 | - | 10 - 11 | 1562 | 78 | ||
AK158552.1 | - | 10 - 11 | 1502 | 78 | ||
BC004848.1 | - | 10 - 11 | 1498 | 78 | ||
BC024835.1 | - | 7 - 8 | 1001 | 78 | ||
BC042502.1 | - | 10 - 11 | 1479 | 5005209 | 78 | |
S50213.1 | - | 9 - 10 | 1376 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |