Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tshr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113404.1 | NP_001106875.1 | 5 - 6 | 560 | 138 | |
NM_011648.5 | NP_035778.3 | 5 - 6 | 560 | 138 | ||
GenBank mRNA | AK014519.1 | - | 5 - 6 | 559 | 138 | |
AK021020.1 | - | 5 - 6 | 554 | 138 | ||
AK029084.1 | - | 5 - 6 | 526 | 138 | ||
AK041086.1 | - | 5 - 6 | 537 | 138 | ||
BC028754.1 | - | 5 - 6 | 515 | 138 | ||
BC086691.1 | - | 5 - 6 | 489 | 30682686 | 138 | |
BC092523.1 | - | 5 - 6 | 502 | 30650002 | 138 | |
U02601.1 | - | 5 - 6 | 542 | 138 | ||
U02602.1 | - | 5 - 6 | 542 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |