Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199310.2 | NP_955014.2 | 2 - 3 | 324 | 100 | |
GenBank mRNA | AK040670.1 | - | 2 - 3 | 294 | 100 | |
AK132604.1 | - | 2 - 3 | 316 | 100 | ||
AK133813.1 | - | 2 - 3 | 298 | 100 | ||
AK147152.1 | - | 2 - 3 | 300 | 100 | ||
BC023926.1 | - | 2 - 3 | 318 | 5324154 | 100 | |
BY262121.1 | - | 2 - 3 | 330 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |